Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
round carboy, with handles
Products
Product Category
carboys (8)
Brand
Sigma-Aldrich (1)
Manufacturer
Diameter (mm)
Height (mm)
Color
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
C5192
Nalgene
®
round carboy, with handles,
nominal capacity 20 L (5.5 gal), polypropylene
Expand
View Pricing
C4442
Nalgene
®
round carboy, with handles,
nominal capacity 10 L (2.5 gal), polypropylene
Expand
View Pricing
C2692
Nalgene
®
round carboy, with handles,
nominal capacity 10 L (2.5 gal), low-density polyethylene
Expand
View Pricing
C4317
Nalgene
®
round carboy, with handles,
nominal capacity 50 L (13 gal), low-density polyethylene
Expand
View Pricing
C3817
Nalgene
®
round carboy, with handles,
nominal capacity 20 L (5.5 gal), low-density polyethylene
Expand
View Pricing
C5567
Nalgene
®
round carboy, with handles,
nominal capacity 50 L (13 gal), polypropylene
Expand
View Pricing
C2817
Nalgene
®
round carboy, with handles,
nominal capacity 15 L (4 gal), low-density polyethylene
Expand
View Pricing
C4192
Nalgene
®
round carboy, with handles,
nominal capacity 25 L (6.5 gal), low-density polyethylene
Expand
View Pricing