Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
Polymethylpentene Erlenmeyer flask
Products
Product Category
flasks (4)
Brand
Sigma-Aldrich (1)
Manufacturer
Capacity (mL)
Cap
Closure Type
Color
Package Size
Sterility
Style
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z362794
Nalgene
®
Polymethylpentene Erlenmeyer flask,
capacity 125 mL
Expand
View Pricing
Z362808
Nalgene
®
Polymethylpentene Erlenmeyer flask,
capacity 250 mL
Expand
View Pricing
Z362816
Nalgene
®
Polymethylpentene Erlenmeyer flask,
capacity 500 mL
Expand
View Pricing
Z362824
Nalgene
®
Polymethylpentene Erlenmeyer flask,
capacity 1,000 mL
Expand
View Pricing