Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
PassPort
™
IP2 bottles
Products
Product Category
bottles (9)
Brand
Sigma-Aldrich (7)
Capacity (mL)
Diameter (mm)
Cap
Color
Vial Bottle Color
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z261084
Nalgene
®
PassPort
™
IP2 bottles,
Narrow-mouth, capacity 125 mL
Expand
View Pricing
Z261068
Nalgene
®
PassPort
™
IP2 bottles,
Narrow-mouth, capacity 30 mL
Expand
View Pricing
Z261076
Nalgene
®
PassPort
™
IP2 bottles,
Narrow-mouth, capacity 60 mL
Expand
View Pricing
Z261157
Nalgene
®
PassPort
™
IP2 bottles,
Wide-mouth, capacity 500 mL
Expand
View Pricing
Z261130
Nalgene
®
PassPort
™
IP2 bottles,
Wide-mouth, capacity 125 mL
Expand
View Pricing
Z261106
Nalgene
®
PassPort
™
IP2 bottles,
Narrow-mouth, capacity 500 mL
Expand
View Pricing
Z261114
Nalgene
®
PassPort
™
IP2 bottles,
Narrow-mouth, capacity 1,000 mL
Expand
View Pricing
Z261092
Nalgene
®
PassPort
™
IP2 bottles,
Narrow-mouth, capacity 250 mL
Expand
View Pricing
Z261149
Nalgene
®
PassPort
™
IP2 bottles,
Wide-mouth, capacity 250 mL
Expand
View Pricing