Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
narrow-mouth bottle
Products
Product Category
bottles (18)
Manufacturer
Capacity (mL)
Volume (ml)
Color
Package Size
Special Grade
Natural (12)
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
TMO312089-0032
Nalgene
®
narrow-mouth bottle,
natural high-density polyethylene, volume 1000 mL, case of 50 ea
Expand
View Pricing
TMO312089-0016
Nalgene
®
narrow-mouth bottle,
natural high-density polyethylene, size 500 mL, case of 125 ea
Expand
View Pricing
TMO312089-0004
Nalgene
®
narrow-mouth bottle,
natural high-density polyethylene, size 125 mL, case of 500 ea
Expand
View Pricing
TMO312085-0004
Nalgene
®
narrow-mouth bottle,
amber high-density polyethylene, volume 125 mL, case of 500 ea
Expand
View Pricing
TMO312089-0008
Nalgene
®
narrow-mouth bottle,
natural high-density polyethylene, size 250 mL, case of 250 ea
Expand
View Pricing
TMO312089-0002
Nalgene
®
narrow-mouth bottle,
natural high-density polyethylene, size 60 mL, case of 1000 ea
Expand
View Pricing
TMO312085-0032
Nalgene
®
narrow-mouth bottle,
amber high-density polyethylene, volume 1000 mL, case of 50 ea
Expand
View Pricing
TMO312087-0001
Nalgene
®
narrow-mouth bottle,
natural polypropylene copolymer, size 30 mL, case of 1000 ea
Expand
View Pricing
TMO312087-0008
Nalgene
®
narrow-mouth bottle,
natural polypropylene copolymer, size 250 mL, case of 250 ea
Expand
View Pricing
TMO312087-0016
Nalgene
®
narrow-mouth bottle,
natural polypropylene copolymer, size 500 mL, case of 125 ea
Expand
View Pricing
TMO312085-0008
Nalgene
®
narrow-mouth bottle,
amber high-density polyethylene, size 250 mL, case of 250 ea
Expand
View Pricing
TMO312085-0002
Nalgene
®
narrow-mouth bottle,
amber high-density polyethylene, size 60 mL, case of 1000 ea
Expand
View Pricing
TMO312085-0001
Nalgene
®
narrow-mouth bottle,
amber high-density polyethylene, size 30 mL, case of 1000 ea
Expand
View Pricing
TMO312087-0004
Nalgene
®
narrow-mouth bottle,
natural polypropylene copolymer, size 125 mL, case of 500 ea
Expand
View Pricing
TMO312089-0001
Nalgene
®
narrow-mouth bottle,
natural high-density polyethylene, size 30 mL, case of 1000 ea
Expand
View Pricing
TMO312087-0032
Nalgene
®
narrow-mouth bottle,
natural polypropylene copolymer, size 1000 mL, case of 50 ea
Expand
View Pricing
TMO312087-0002
Nalgene
®
narrow-mouth bottle,
natural polypropylene copolymer, size 60 mL, case of 1000 ea
Expand
View Pricing
TMO312085-0016
Nalgene
®
narrow-mouth bottle,
amber high-density polyethylene, size 500 mL, case of 125 ea
Expand
View Pricing