Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
FEP bottle
Products
Product Category
bottles (11)
Brand
Sigma-Aldrich (2)
Color
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z130230
Nalgene
®
FEP bottle,
capacity 30 mL, Narrow-mouth, clear Bottle
Expand
View Pricing
Z266167
Nalgene
®
FEP bottle,
capacity 1000 mL, Narrow-mouth, clear Bottle
Expand
View Pricing
Z130389
Nalgene
®
FEP bottle,
capacity 250 mL, Wide-mouth, clear Bottle
Expand
View Pricing
Z130370
Nalgene
®
FEP bottle,
capacity 125 mL, Wide-mouth, clear Bottle
Expand
View Pricing
Z130400
Nalgene
®
FEP bottle,
Wide-mouth, capacity 1000 mL, clear Bottle
Expand
View Pricing
Z130397
Nalgene
®
FEP bottle,
Wide-mouth, capacity 500 mL, clear Bottle
Expand
View Pricing
Z130265
Nalgene
®
FEP bottle,
Narrow-mouth, capacity 125 mL, clear Bottle
Expand
View Pricing
Z130303
Nalgene
®
FEP bottle,
Narrow-mouth, capacity 250 mL, clear Bottle
Expand
View Pricing
Z266175
Nalgene
®
FEP bottle,
capacity 2000 mL, Narrow-mouth, clear Bottle
Expand
View Pricing
Z130311
Nalgene
®
FEP bottle,
Narrow-mouth, capacity 500 mL, clear Bottle
Expand
View Pricing
Z130249
Nalgene
®
FEP bottle,
Narrow-mouth, capacity 60 mL, clear Bottle
Expand
View Pricing