Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
dropper bottle with screw closure
Products
Product Category
bottles (10)
Brand
Sigma-Aldrich (4)
Color
Capacity (mL)
Color
Vial Bottle Color
Package Size
Special Grade
Natural (2)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
B6055
Nalgene
®
dropper bottle with screw closure,
capacity 4 mL, colorless bottle, white cap
Expand
View Pricing
B6805
Nalgene
®
dropper bottle with screw closure,
capacity 4 mL, white bottle, assorted colors cap
Expand
View Pricing
B6555
Nalgene
®
dropper bottle with screw closure,
capacity 8 mL, white bottle, white cap
Expand
View Pricing
B7055
Nalgene
®
dropper bottle with screw closure,
capacity 15 mL, natural bottle, assorted colors cap
Expand
View Pricing
B6305
Nalgene
®
dropper bottle with screw closure,
capacity 15 mL, colorless bottle, white cap
Expand
View Pricing
B7430
Nalgene
®
dropper bottle with screw closure,
capacity 15 mL, white bottle, assorted colors cap
Expand
View Pricing
B6930
Nalgene
®
dropper bottle with screw closure,
capacity 8 mL, natural bottle, assorted colors cap
Expand
View Pricing
B6680
Nalgene
®
dropper bottle with screw closure,
capacity 15 mL, white bottle, white cap
Expand
View Pricing
B6180
Nalgene
®
dropper bottle with screw closure,
capacity 8 mL, colorless bottle, white cap
Expand
View Pricing
B6430
Nalgene
®
dropper bottle with screw closure,
capacity 4 mL, white bottle, white cap
Expand
View Pricing