Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
-Acetyl-D-glucosamine
Synonym(s):
2-Acetamido-2-deoxy-D-glucose, D-GlcNAc
Empirical Formula (Hill Notation):
C
8
H
15
NO
6
CAS Number:
7512-17-6
Molecular Weight:
221.21
EC Number:
231-368-2
Products
Product Category
carbohydrates (3)
monosaccharides (2)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Tissue
Quality Segment
Optical Isomer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A8625
N
-Acetyl-D-glucosamine,
≥99% (HPLC)
Expand
View Pricing
A3286
N
-Acetyl-D-glucosamine,
BioReagent, suitable for cell culture
Expand
View Pricing
A4106
N
-Acetyl-D-glucosamine,
≥95% (HPLC)
Expand
View Pricing