Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Pure.Amp
™
qPCR 8 Tube Strips
Products
Product Category
tubes (3)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
MTCP3802Q
MTC
™
Bio Pure.Amp
™
qPCR 8 Tube Strips,
capacity 0.2 mL, clear tube, clear cap (optical), flat cap
Expand
View Pricing
MTCP3801Q
MTC
™
Bio Pure.Amp
™
qPCR 8 Tube Strips,
capacity 0.1 mL, clear tube, clear cap (optical), flat cap
Expand
View Pricing
MTCP3801QW
MTC
™
Bio Pure.Amp
™
qPCR 8 Tube Strips,
capacity 0.1 mL, white tube, separate, optical clear flat cap
Expand
View Pricing
MTCP3802QW
MTC
™
Bio Pure.Amp
™
qPCR 8 Tube Strips,
capacity 0.2 mL, white tube, clear cap (optical), flat cap
Expand
View Pricing
MTCP3801QF
MTC
™
Bio Pure.Amp
™
qPCR 8 Tube Strips,
capacity 0.1 mL, tube frosted, separate, optical clear flat caps
Expand
View Pricing