Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid
Products
Product Category
racks (16)
Manufacturer
Height (mm)
Length (mm)
Color
Package Size
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
MTCR2781A
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, assorted colors, to hold, 81 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2700A
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, assorted colors, to hold, 100 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2781
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, white, to hold, 81 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2700
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, white, to hold, 100 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR3700
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, white, to hold, 100 x cryovials, pack of 50 ea
Expand
View Pricing
MTCR3781
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, white, to hold, 81 x cryovials, pack of 50 ea
Expand
View Pricing
MTCR2781P
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, purple, to hold, 81 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2700B
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, blue, to hold, 100 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2781R
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, red, to hold, 81 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2781G
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, green, to hold, 81 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2781B
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, blue, to hold, 81 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2700G
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, green, to hold, 100 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2781O
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, orange, to hold, 81 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2700O
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, orange, to hold, 100 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2700R
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, red, to hold, 100 x cryovials, pkg of 5 ea
Expand
View Pricing
MTCR2700P
MTC
™
Bio FlipTop
™
Freezer Box With Hinged Lid,
cardboard, purple, to hold, 100 x cryovials, pkg of 5 ea
Expand
View Pricing