Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Autoclave Bags
Products
Product Category
bags (5)
Manufacturer
Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
MTCA9004R
MTC
™
Bio Autoclave Bags,
8.5in. x 11in. (21.6 x 27.9cm), red (black printed warnings)
Expand
View Pricing
MTCA9000C
MTC
™
Bio Autoclave Bags,
size 31-66 cm (12-26 in.), clear (blue printed warnings)
Expand
View Pricing
MTCA9002R
MTC
™
Bio Autoclave Bags,
24 in. x 32 in. (61 x 81 cm), red (black printed warnings)
Expand
View Pricing
MTCA9002C
MTC
™
Bio Autoclave Bags,
24 in. x 32 in. (61 x 81 cm), clear (blue printed warnings)
Expand
View Pricing
MTCA9000R
MTC
™
Bio Autoclave Bags,
W × H 12 in. (31 cm) × 26 in. (66 cm), red (black printed warnings), pack of 200 ea (bulk bag)
Expand
View Pricing