
Moxidectin

Synonym(s):
Moxidectin
Empirical Formula (Hill Notation):
C37H53NO8
CAS Number:
Molecular Weight:
639.82
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing