Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Molybdenum(IV) sulfide
Synonym(s):
Molybdenum disulfide
Empirical Formula (Hill Notation):
MoS
2
CAS Number:
1317-33-5
Molecular Weight:
160.07
EC Number:
215-263-9
Products
Brand
Sigma-Aldrich (3)
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Application
Particle Size (µm)
Special Grade
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
69860
Molybdenum(IV) sulfide,
powder
Expand
View Pricing
234842
Molybdenum(IV) sulfide,
powder, <2 μm, 98%
Expand
View Pricing
804169
Molybdenum(IV) sulfide,
nanopowder, 90 nm diameter (APS), 99% trace metals basis
Expand
View Pricing