Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mitomycin
Synonym(s):
Mitomycin C
Empirical Formula (Hill Notation):
C
15
H
18
N
4
O
5
CAS Number:
50-07-7
Molecular Weight:
334.33
EC Number:
200-008-6
Products
Product Category
antibiotics (1)
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1444707
Mitomycin,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
Y0000378
Mitomycin,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing