Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microtubes
Products
Product Category
tubes (27)
Color
Manufacturer
Sterility
Filter Products
Showing 1-20 of 27
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
AXYMCT150CS
Microtubes,
size 1.5 mL, clear, pkg of 10x250tubes/cs, sterile
Expand
View Pricing
BR780555
Microtubes,
polypropylene, 5 mL
Expand
View Pricing
AXYMCT200CS
Microtubes,
size 2.0 mL, clear, pkg of 10x250tubes/cs, sterile
Expand
View Pricing
AXYMCT175C
Microtubes,
size 1.7 mL, clear, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT200SP
Microtubes,
size 2.0 mL, in spectrum pack, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT150NC
Microtubes,
no cap, size 1.5 mL, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT175CS
Microtubes,
size 1.7 mL, clear, pkg of 10x250tubes/cs, sterile
Expand
View Pricing
AXYMCT060CS
Microtubes,
size 0.6 mL, clear, pkg of 10x500tubes/cs, sterile
Expand
View Pricing
AXYMCT060C
Microtubes,
size 0.6 mL, clear, pkg of 10x1000tubes/cs
Expand
View Pricing
AXYMCT200NC
Microtubes,
no cap, size 2.0 mL, clear, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT060X
Microtubes,
size 0.6 mL, amber, pkg of 10x1000tubes/cs
Expand
View Pricing
AXYMCT175X
Microtubes,
size 1.7 mL, amber, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT175A
Microtubes,
size 1.7 mL, assorted colors, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT060O
Microtubes,
size 0.6 mL, orange, pkg of 10x1000tubes/cs
Expand
View Pricing
AXYMCT060A
Microtubes,
size 0.6 mL, assorted colors, pkg of 10x1000tubes/cs
Expand
View Pricing
AXYMCT175G
Microtubes,
size 1.7 mL, green, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT175SP
Microtubes,
size 1.7 mL, in spectrum pack, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT060V
Microtubes,
size 0.6 mL, violet, pkg of 10x1000tubes/cs
Expand
View Pricing
AXYMCT175R
Microtubes,
size 1.7 mL, red, pkg of 10x500tubes/cs
Expand
View Pricing
AXYMCT060R
Microtubes,
size 0.6 mL, red, pkg of 10x1000tubes/cs
Expand
View Pricing
Page 1 of 2
Page 1 of 2