Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microfil
®
with EZ-Pak
®
Products
Product Category
funnels (6)
Brand
Millipore (8)
Agency
Capacity (mL)
Diameter (mm)
Compatibility
Hydrophobicity
Application
Color
Pore size (µm)
Sterility
Sterilization Method
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
MZHAWG101
Microfil
®
with EZ-Pak
®
,
pore size 0.45 μm
Expand
View Pricing
MZHAWG251
Microfil
®
with EZ-Pak
®
,
ready-to-use, pore size 0.45 μm, mixed cellulose esters (MCE) filter, sample volume 250 mL, filter diam. 47 mm
Expand
View Pricing
MZHABG101
Microfil
®
with EZ-Pak
®
,
ready-to-use, sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, black filter, gridded filter
Expand
View Pricing
MZHABG251
Microfil
®
with EZ-Pak
®
,
pore size 0.45 μm
Expand
View Pricing
MZGSWG101
Microfil
®
with EZ-Pak
®
,
pore size 0.22 μm
Expand
View Pricing
MZAABG101
Microfil
®
with EZ-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, black filter, gridded filter
Expand
View Pricing
MZAAWG251
Microfil
®
with EZ-Pak
®
,
sample volume 250 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, white filter, gridded filter
Expand
View Pricing
MZAAWG101
Microfil
®
with EZ-Pak
®
,
sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, white filter, gridded filter
Expand
View Pricing