Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler Toledo SevenExcellence
™
pH/Ion meter S500
Products
Manufacturer
pH Value
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
MT30046249
Mettler Toledo SevenExcellence
™
pH/Ion meter S500,
S500 standard kit, with InLab
®
Expert Pro-ISM, AC/DC input 230 V AC, universal plug set
Expand
View Pricing
MT30046251
Mettler Toledo SevenExcellence
™
pH/Ion meter S500,
pH/Ion meter S500 fluoride kit, with perfectION
™
Fluoride, AC/DC input 230 V AC, universal plug set
Expand
View Pricing
MT30046250
Mettler Toledo SevenExcellence
™
pH/Ion meter S500,
S500 biotechnology kit, with InLab
®
Routine Pro-ISM cable kit, AC/DC input 230 V AC, universal plug set
Expand
View Pricing
MT30046248
Mettler Toledo SevenExcellence
™
pH/Ion meter S500,
S500 basic (no electrode), AC/DC input 230 V AC, universal plug set
Expand
View Pricing