Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler-Toledo Calibration substance ME 30130598, Methyltriphenylphosphoniumbromide
Synonym(s):
Methyltriphenylphosphonium bromide
Linear Formula:
CH
3
P(C
6
H
5
)
3
Br
CAS Number:
1779-49-3
Molecular Weight:
357.22
EC Number:
217-218-9
Products
Brand
Supelco (1)
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
94993
Mettler-Toledo Calibration substance ME 30130598, Methyltriphenylphosphoniumbromide,
tracable to primary standards (LGC)
Expand
View Pricing