Skip to Content
Merck
Metoprolol succinate Pharmaceutical Secondary Standard; Certified Reference Material

Metoprolol succinate

Metoprolol succinate Pharmaceutical Secondary Standard; Certified Reference Material
Empirical Formula (Hill Notation):
C30H50N2O6 · C4H6O4
CAS Number:
Molecular Weight:
652.82

Products

Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade