
Metoprolol fumarate

Synonym(s):
Metoprolol fumarate
Empirical Formula (Hill Notation):
C30H50N2O6 · C4H4O4
CAS Number:
Molecular Weight:
650.80
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2