Skip to Content
Merck
Methyl 6-methylpyridine-3-carboxylate 97%

Methyl 6-methylpyridine-3-carboxylate

Methyl 6-methylpyridine-3-carboxylate 97%
Synonym(s):
Methyl 6-methylnicotinate
Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing