Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methoxypolyethylene glycol maleimide
Synonym(s):
MeO-PEG-Mal, PEG-maleimide, mono-Methyl polyethylene glycol 2-maleimidoethyl ether
CAS Number:
99126-64-4
Products
Product Category
detergents (1)
Brand
Sigma-Aldrich (3)
Physical Form
Purity
Quality Segment
Functional Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
712469
Methoxypolyethylene glycol maleimide,
PEG average M
n
10,000 g/mol
Expand
View Pricing
712558
Methoxypolyethylene glycol maleimide,
750
Expand
View Pricing
63187
Methoxypolyethylene glycol maleimide,
≥90% (NMR), 5,000
Expand
View Pricing