Methadone Hydrochloride
Synonym(s):
(±)-Methadone hydrochloride, 6-Dimethylamino-4,4-diphenylheptan-3-one hydrochloride
Empirical Formula (Hill Notation):
C21H27NO · HCl
CAS Number:
Molecular Weight:
345.91
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2