Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Metal Packed GC Column (General Configuration)
Products
Product Category
columns (26)
Brand
Supelco (20)
Manufacturer
Column Type
ID (mm)
Length (m)
Compatibility
Column Material
Technique
Filter Products
Showing 1-20 of 26
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
12390-U
Metal Packed GC Column (General Configuration),
phase none, matrix 60/80 Carboxen-1000 support, L × O.D. × I.D. 15.0 ft (4.6 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13052-U
Metal Packed GC Column (General Configuration),
phase none, matrix 80/100 Porapak N support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13047-U
Metal Packed GC Column (General Configuration),
phase none, matrix 45/60 Molecular Sieve 13X support, L × O.D. × I.D. 3.0 ft (0.91 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12380
Metal Packed GC Column (General Configuration),
phase none, matrix 40/60 Carboxen-1000 support, L × O.D. × I.D. 5.0 ft (1.5 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12437
Metal Packed GC Column (General Configuration),
phase none, matrix 80/100 Porapak Q support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13979-U
Metal Packed GC Column (General Configuration),
phase none, matrix 80/100 Porapak Q support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13801
Metal Packed GC Column (General Configuration),
phase none, matrix 80/100 HayeSep Q support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13130-U
Metal Packed GC Column (General Configuration),
phase none, matrix 45/60 Molecular Sieve 5A support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13133-U
Metal Packed GC Column (General Configuration),
phase none, matrix 60/80 Molecular Sieve 5A support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12212
Metal Packed GC Column (General Configuration),
phase 10% Carbowax 20M, matrix 80/100 Chromosorb W AW support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13036-U
Metal Packed GC Column (General Configuration),
phase none, matrix 45/60 Molecular Sieve 13X support, L × O.D. × I.D. 10.0 ft (3.0 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12809-U
Metal Packed GC Column (General Configuration),
phase 23% SP
®
-1700, matrix 80/100 Chromosorb P AW support, L × O.D. × I.D. 30.0 ft (9.1 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12449
Metal Packed GC Column (General Configuration),
phase 3% Petrocol B, matrix 80/100 SUPELCOPORT support, L × O.D. × I.D. 1.7 ft (0.52 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12873
Metal Packed GC Column (General Configuration),
phase 20% TCEP, matrix 80/100 Chromosorb P AW support, L × O.D. × I.D. 1.8 ft (0.55 m) × 1/16 in. × 0.75 mm
Expand
View Pricing
13128-U
Metal Packed GC Column (General Configuration),
phase 5% Carbowax 20M, matrix 80/100 Chromosorb W HP support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12719
Metal Packed GC Column (General Configuration),
phase SP
®
-1000, 10%, matrix 80/100 SUPELCOPORT support, L × O.D. × I.D. 20.0 ft (6.1 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12429
Metal Packed GC Column (General Configuration),
phase 10% SP-2100, matrix 80/100 SUPELCOPORT support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
12370-U
Metal Packed GC Column (General Configuration),
phase none, matrix 40/60 Carboxen-1000 support, L × O.D. × I.D. 2.0 ft (0.61 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13067-U
Metal Packed GC Column (General Configuration),
phase none, matrix 80/100 HayeSep N support, L × O.D. × I.D. 8.0 ft (2.4 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
13860-U
Metal Packed GC Column (General Configuration),
phase 0.2% Carbowax 1500, matrix 60/80 Carbopack C support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2