Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mesitylene
Synonym(s):
1,3,5-Trimethylbenzene
Linear Formula:
C
6
H
3
(CH
3
)
3
CAS Number:
108-67-8
Molecular Weight:
120.19
EC Number:
203-604-4
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (3)
Supelco (2)
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
certified reference material (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
M7200
Mesitylene,
98%
Expand
View Pricing
8.05890
Mesitylene,
Technipur
®
, for synthesis
Expand
View Pricing
140864
Mesitylene,
reagent grade, 97%
Expand
View Pricing
63908
Mesitylene,
analytical standard
Expand
View Pricing
61927
Mesitylene,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing