Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MarvelX
™
UHPLC Connection Systems
Products
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
51888-U
MarvelX
™
UHPLC Connection Systems,
PEEK Lined Stainless Steel Tubing, I.D. × L 100 μm × 350 mm
Expand
View Pricing
51896-U
MarvelX
™
UHPLC Connection Systems,
Stainless Steel Tubing, I.D. × L 125 μm × 350 mm
Expand
View Pricing
51894-U
MarvelX
™
UHPLC Connection Systems,
Stainless Steel Tubing, I.D. 125 μm × 150 mm
Expand
View Pricing
51882-U
MarvelX
™
UHPLC Connection Systems,
PEEK Lined Stainless Steel Tubing, I.D. × L 25 μm × 350 mm
Expand
View Pricing
51875-U
MarvelX
™
UHPLC Connection Systems,
PEEK Lined Stainless Steel Tubing, I.D. × L 100 μm × 150 mm
Expand
View Pricing
51874-U
MarvelX
™
UHPLC Connection Systems,
PEEK Lined Stainless Steel Tubing, I.D. × L 25 μm × 150 mm
Expand
View Pricing
51897-U
MarvelX
™
UHPLC Connection Systems,
Stainless Steel Tubing, I.D. × L 254 μm × 150 mm
Expand
View Pricing
51899-U
MarvelX
™
UHPLC Connection Systems,
Stainless Steel Tubing, I.D. × L 254 μm × 350 mm
Expand
View Pricing