Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Maleimide-poly
(
ethylene glycol)-
b-
poly(ε-caprolactone)
Linear Formula:
C
7
H
7
N
2
O
3
[C
2
H
4
O]
n
[C
6
H
10
O
2
]
m
H
Products
Brand
Sigma-Aldrich (2)
Color
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
902381
Maleimide-poly
(
ethylene glycol)-
b-
poly(ε-caprolactone),
PEG average M
n
5000, PCL average M
n
5000
Expand
View Pricing
915343
Maleimide-poly
(
ethylene glycol)-
b-
poly(ε-caprolactone),
PEG average M
n
5,000, PCL average M
n
10,000
Expand
View Pricing