
Maleic acid

Synonym(s):
cis-Butenedioic acid, Toxilic acid
Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
pK Value
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing