Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnetic copper
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF11423708
Magnetic copper,
wire, Cu 99.96%/Fe 0.04%, 0.5 mm diameter, length 2 m, filament diameter 0.1 microns
Expand
View Pricing
GF25090916
Magnetic copper,
wire, Cu 99.96%/Fe 0.04%, 0.5 mm diameter, length 20 m, filament diameter 0.1 microns
Expand
View Pricing
GF36364743
Magnetic copper,
wire, Cu 99.96%/Fe 0.04%, 0.5 mm diameter, length 50 m, filament diameter 0.1 microns
Expand
View Pricing
GF27963209
Magnetic copper,
wire, Cu 99.96%/Fe 0.04%, 0.5 mm diameter, length 5 m, filament diameter 0.1 microns
Expand
View Pricing
GF52091388
Magnetic copper,
wire, Cu 99.96%/Fe 0.04%, 0.5 mm diameter, length 1 m, filament diameter 0.1 microns
Expand
View Pricing
GF95400727
Magnetic copper,
wire, Cu99.96%/Fe 0.04%, 0.5 mm diameter, filament diameter 0.1 microns, length 10 m
Expand
View Pricing