Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
GF10546398
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
490mm rod, 3mm diameter, extruded
Expand
View Pricing
GF16361748
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
500mm rod, 25mm diameter, extruded
Expand
View Pricing
GF67837281
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 200 x 200mm, 0.25mm thickness
Expand
View Pricing
GF99246561
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
980mm rod, 3mm diameter, extruded
Expand
View Pricing
GF65085677
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
100mm rod, 25mm diameter, extruded
Expand
View Pricing
GF04221841
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
1000mm rod, 25mm diameter, extruded
Expand
View Pricing
GF74847965
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 200 x 500mm, 2.0mm thickness
Expand
View Pricing
GF56611290
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 100 x 100mm, 0.25mm thickness
Expand
View Pricing
GF21221895
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 100 x 100mm, 2.0mm thickness
Expand
View Pricing
GF93367974
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 200 x 500mm, 1.0mm thickness
Expand
View Pricing
GF61324242
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
200mm rod, 25mm diameter, extruded
Expand
View Pricing
GF47027112
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 100 x 100mm, 1.0mm thickness
Expand
View Pricing
GF68379160
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 200 x 200mm, 2.0mm thickness
Expand
View Pricing
GF98994762
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 200 x 200mm, 1.0mm thickness
Expand
View Pricing
GF96338357
Magnesium-aluminum-zinc alloy, Mg96Al3Zn1,
foil, 200 x 500mm, 0.25mm thickness
Expand
View Pricing