Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lyophilized Cationic Liposomes
Products
Brand
Sigma-Aldrich (9)
Color
Physical Form
Chemical Composition
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
LDP-501
Lyophilized Cationic Liposomes,
DOTAP:DOPC (50:50 molar ratio)
Expand
View Pricing
LDP-503
Lyophilized Cationic Liposomes,
DOTAP:DOPC (30:70 molar ratio)
Expand
View Pricing
LDP-502
Lyophilized Cationic Liposomes,
DOTAP:DOPC (40:60 molar ratio)
Expand
View Pricing
LDP-508
Lyophilized Cationic Liposomes,
DOTAP:DOPC (1:99 molar ratio)
Expand
View Pricing
LDP-509
Lyophilized Cationic Liposomes,
DOTAP:DOPC (0.5:99.5 molar ratio)
Expand
View Pricing
LDP-504
Lyophilized Cationic Liposomes,
DOTAP:DOPC (20:80 molar ratio)
Expand
View Pricing
LDP-505
Lyophilized Cationic Liposomes,
DOTAP:DOPC (10:90 molar ratio)
Expand
View Pricing
LDP-507
Lyophilized Cationic Liposomes,
DOTAP:DOPC (2:98 molar ratio)
Expand
View Pricing
LDP-506
Lyophilized Cationic Liposomes,
DOTAP:DOPC (5:95 molar ratio)
Expand
View Pricing