Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Luteolin
Synonym(s):
3′,4′,5,7-Tetrahydroxyflavone, Luteolin
Empirical Formula (Hill Notation):
C
15
H
10
O
6
CAS Number:
491-70-3
Molecular Weight:
286.24
EC Number:
207-741-0
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (2)
primary reference standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
L9283
Luteolin,
≥98% (TLC), powder, antioxidant
Expand
View Pricing
440025
Luteolin,
An antioxidant flavonoid and a free radical scavenger.
Expand
View Pricing
PHL89245
Luteolin,
phyproof
®
Reference Substance
Expand
View Pricing
72511
Luteolin,
analytical standard
Expand
View Pricing
03880590
Luteolin,
primary reference standard
Expand
View Pricing