Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Low alloy steel
Products
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
certified reference material (31)
Filter Products
Showing 1-20 of 32
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
NIST361
Low alloy steel,
NIST
®
SRM
®
361, (AISI 4340)
Expand
View Pricing
NIST1766
Low alloy steel,
NIST
®
SRM
®
1766
Expand
View Pricing
NIST1765
Low alloy steel,
NIST
®
SRM
®
1765
Expand
View Pricing
NIST1764A
Low alloy steel,
NIST
®
SRM
®
1764a
Expand
View Pricing
NIST1761A
Low alloy steel,
NIST
®
SRM
®
1761a
Expand
View Pricing
NIST1228
Low alloy steel,
NIST
®
SRM
®
1228, 0.1% C
Expand
View Pricing
NIST1763B
Low alloy steel,
disk form, NIST
®
SRM
®
1763b
Expand
View Pricing
NIST72G
Low alloy steel,
NIST
®
SRM
®
72g, (AISI 4130)
Expand
View Pricing
NIST2160
Low alloy steel,
NIST
®
SRM
®
2160, carbon & sulfur only
Expand
View Pricing
NIST1218
Low alloy steel,
NIST SRM 1218, high silicon
Expand
View Pricing
NIST1227
Low alloy steel,
NIST
®
SRM
®
1227, basic open hearth, 1%C
Expand
View Pricing
NIST1135
Low alloy steel,
NIST
®
SRM
®
1135, high silicon
Expand
View Pricing
NIST1270
Low alloy steel,
NIST
®
SRM
®
1270, Cr-Mo (A336) (F-22)
Expand
View Pricing
NIST2167
Low alloy steel,
NIST
®
SRM
®
2167, G
Expand
View Pricing
NIST2171
Low alloy steel,
NIST
®
SRM
®
2171, (HSLA-100)
Expand
View Pricing
NIST293
Low alloy steel,
NIST
®
SRM
®
293, Cr-Ni-Mo (AISI 8620)
Expand
View Pricing
NISTC1285
Low alloy steel,
NIST
®
SRM
®
C1285, (A242) (mod.)
Expand
View Pricing
NIST1224
Low alloy steel,
NIST
®
SRM
®
1224, carbon (AISI 1078)
Expand
View Pricing
NIST1271
Low alloy steel,
NIST
®
SRM
®
1271, Ni-Cr-Cu-Mo (HSLA-100)
Expand
View Pricing
NIST1225
Low alloy steel,
NIST
®
SRM
®
1225, (AISI 4130)
Expand
View Pricing
Page 1 of 2
Page 1 of 2