Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Lactic Dehydrogenase from bovine heart
Synonym(s):
(
S
)-Lactate: NAD
+
oxidoreductase, L-LDH, LAD, LD
CAS Number:
9001-60-9
EC Number:
232-617-8
Products
Product Category
enzymes (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Manufacturer
Physical Form
Tissue
Application
Optical Isomer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
L2625
L-Lactic Dehydrogenase from bovine heart,
Type III, ammonium sulfate suspension, ≥500 units/mg protein
Expand
View Pricing
L3916
L-Lactic Dehydrogenase from bovine heart,
1000 units/mL
Expand
View Pricing
L1006
L-Lactic Dehydrogenase from bovine heart,
Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Expand
View Pricing