Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium chloride
Synonym(s):
Lithium chloride
Linear Formula:
LiCl
CAS Number:
7447-41-8
Molecular Weight:
42.39
EC Number:
231-212-3
Products
Product Category
salts (11)
Brand
Sigma-Aldrich (12)
Supelco (1)
Sustainability
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Anhydrous (5)
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
310468
Lithium chloride,
ACS reagent, ≥99%
Expand
View Pricing
793620
Lithium chloride,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, 99%
Expand
View Pricing
L9650
Lithium chloride,
for molecular biology, ≥99%
Expand
View Pricing
203637
Lithium chloride,
powder, ≥99.98% trace metals basis
Expand
View Pricing
213233
Lithium chloride,
ReagentPlus
®
, 99%
Expand
View Pricing
746460
Lithium chloride,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
1.05679
Lithium chloride,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
62476
Lithium chloride,
BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
Expand
View Pricing
916013
Lithium chloride,
anhydrous, 99.95% trace metals basis
Expand
View Pricing
429457
Lithium chloride,
AnhydroBeads
™
, −10 mesh, 99.998% trace metals basis
Expand
View Pricing
73036
Lithium chloride,
puriss. p.a., anhydrous, ≥99.0% (AT)
Expand
View Pricing
L4408
Lithium chloride,
BioXtra, ≥99.0% (titration)
Expand
View Pricing
449881
Lithium chloride,
AnhydroBeads
™
, −10 mesh, ≥99.9% trace metals basis
Expand
View Pricing