Skip to Content
Merck
Linalool oxide natural, ≥95%

Linalool oxide

Linalool oxide natural, ≥95%
Synonym(s):
2-(5-Methyl-5-vinyltetrahydro-1-furyl)-2-propanol
Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
EC Number:

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing