Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lead paint film for portable x-ray fluorescence analyzers
Products
Manufacturer
Format
Quality Segment
Application
Special Grade
certified reference material (6)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
NIST2571
Lead paint film for portable x-ray fluorescence analyzers,
NIST
®
SRM
®
2571, nominal 3.5 mg/cm2 (color code: yellow)
Expand
View Pricing
NIST2570
Lead paint film for portable x-ray fluorescence analyzers,
NIST
®
SRM
®
2570, blank (color code: white)
Expand
View Pricing
NIST2574
Lead paint film for portable x-ray fluorescence analyzers,
NIST
®
SRM
®
2574, nominal 0.7 mg/cm2 (color code: gold)
Expand
View Pricing
NIST2572
Lead paint film for portable x-ray fluorescence analyzers,
NIST
®
SRM
®
2572, nominal 1.6 mg/cm2 (color code: orange)
Expand
View Pricing
NIST2575
Lead paint film for portable x-ray fluorescence analyzers,
NIST
®
SRM
®
2575, nominal 0.3 mg/cm2 (color code: green)
Expand
View Pricing
NIST2576
Lead paint film for portable x-ray fluorescence analyzers,
NIST
®
SRM
®
2576, nominal 5.6 mg/cm2 (color code: blue)
Expand
View Pricing