Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Amino Acid Oxidase from
Crotalus adamanteus
Synonym(s):
L-AAO, L-Amino acid: oxygen oxidoreductase (deaminating), L-Amino acid:oxygen oxidoreductase (deaminating)
CAS Number:
9000-89-9
EC Number:
232-564-0
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (2)
Biological Source
Physical Form
Quality Segment
Optical Isomer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
A9253
L-Amino Acid Oxidase from
Crotalus adamanteus
,
Type I (dried venom)
Expand
View Pricing
A9378
L-Amino Acid Oxidase from
Crotalus adamanteus
,
Type IV, ≥4.0 units/mg protein, aqueous suspension
Expand
View Pricing