Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Laboratory brushes
Products
Length (in)
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z281344
Laboratory brushes,
Flask, L 12 in., brush diam. × L 1 3/8 in. × 3 1/2 in.
Expand
View Pricing
Z281255
Laboratory brushes,
Test tube, L 9 in., brush diam. × L 3/4 in. × 3 1/4 in.
Expand
View Pricing
Z281247
Laboratory brushes,
Micro tube, L 7 in., brush diam. × L 3/16- 1/2 in. × 3 in.
Expand
View Pricing
Z281301
Laboratory brushes,
Narrow tube, L 12 in., brush diam. × L 1/2 in. × 3 in.
Expand
View Pricing
Z281263
Laboratory brushes,
Test tube, L 10 in., brush diam. × L 1 3/8 in. × 4 in.
Expand
View Pricing
Z281271
Laboratory brushes,
Cylinder, L 11 in., brush diam. × L 2 3/8 in. × 5 in.
Expand
View Pricing
Z281360
Laboratory brushes,
Burette, L 30 in., brush diam. × L 1/2 in. × 3 1/4 in.
Expand
View Pricing
Z281298
Laboratory brushes,
Cylinder, L 20 in., brush diam. × L 2 3/8 in. × 6 in.
Expand
View Pricing
Z281352
Laboratory brushes,
Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
Expand
View Pricing
Z281328
Laboratory brushes,
Pipette, L 17 in., brush diam. × L 3/16- 3/4 in. × 7 1/2 in.
Expand
View Pricing
Z281336
Laboratory brushes,
Funnel, L 13 in., brush diam. × L 2 1/4 in. × 6 in.
Expand
View Pricing
Z281379
Laboratory brushes,
Burette, L 36 in., brush diam. × L 3/4 in. × 3 1/4 in.
Expand
View Pricing