Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Labeling tape
Products
Product Category
labels (10)
Manufacturer
Length (in)
Color
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z768294
Labeling tape,
white, W × L 3/4 in. × 500 in.
Expand
View Pricing
Z768308
Labeling tape,
yellow, W × L 3/4 in. × 500 in.
Expand
View Pricing
Z768324
Labeling tape,
blue, W × L 3/4 in. × 500 in.
Expand
View Pricing
Z768316
Labeling tape,
green, W × L 3/4 in. × 500 in.
Expand
View Pricing
Z768332
Labeling tape,
W × L 3/4 in. × 500 in., red
Expand
View Pricing
Z768359
Labeling tape,
yellow, W × L 3/4 in. × 2160 in.
Expand
View Pricing
Z768340
Labeling tape,
white, W × L 3/4 in. × 2160 in.
Expand
View Pricing
Z768383
Labeling tape,
red, W × L 3/4 in. × 2160 in.
Expand
View Pricing
Z768367
Labeling tape,
green, W × L 3/4 in. × 2160 in.
Expand
View Pricing
Z768375
Labeling tape,
blue, W × L 3/4 in. × 2160 in.
Expand
View Pricing