Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KIMBLE
®
Solid Cap
Products
Product Category
caps (12)
Glassware Class
Color
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
DWK73802-18400
KIMBLE
®
Solid Cap,
18-400 thread, black phenolic cap (with rubber liner)
Expand
View Pricing
DWK73800-15415
KIMBLE
®
Solid Cap,
with rubber liner, phenolic, cap for 15-415
Expand
View Pricing
DWK45066C-15
KIMBLE
®
Solid Cap,
black phenolic cap (with PTFE Liner), cap for 15-415
Expand
View Pricing
DWK73800-13415
KIMBLE
®
Solid Cap,
black phenolic cap, cap for 13-425
Expand
View Pricing
DWK73802-22400
KIMBLE
®
Solid Cap,
black phenolic cap (with rubber liner), 22-400 thread
Expand
View Pricing
DWK45066C-13
KIMBLE
®
Solid Cap,
black phenolic cap (with PTFE Liner), cap for 13-415
Expand
View Pricing
DWK45066C-18
KIMBLE
®
Solid Cap,
black phenolic cap (with PTFE liner), cap for 18-415
Expand
View Pricing
DWK73802-8425
KIMBLE
®
Solid Cap,
black phenolic cap (with rubber liner), cap for 8-425
Expand
View Pricing
DWK45066C-24
KIMBLE
®
Solid Cap,
black phenolic cap (with PTFE Liner), cap for 24-410
Expand
View Pricing
DWK45066C-38
KIMBLE
®
Solid Cap,
black phenolic cap (with PTFE Liner), cap for 38-430
Expand
View Pricing
DWK73802-33430
KIMBLE
®
Solid Cap,
black phenolic cap (with rubber liner), cap for 33-400
Expand
View Pricing
DWK73802-38430
KIMBLE
®
Solid Cap,
black phenolic cap (with rubber liner), cap for 38-400
Expand
View Pricing