Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KIMBLE
®
KIMAX
®
Media bottle
Products
Product Category
bottles (14)
Capacity (mL)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Sterility
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
DWK14395-5000
KIMBLE
®
KIMAX
®
Media bottle,
with cap, round transparent borosilicate glass bottle, bottle capacity (5,000 mL), non-sterile
Expand
View Pricing
DWK14395-10000
KIMBLE
®
KIMAX
®
Media bottle,
with cap, round transparent borosilicate glass bottle, bottle capacity (10,000 mL), non-sterile
Expand
View Pricing
DWK14395-2000
KIMBLE
®
KIMAX
®
Media bottle,
with cap, round transparent borosilicate glass bottle, bottle capacity (2,000 mL), non-sterile
Expand
View Pricing
DWK14395-500
KIMBLE
®
KIMAX
®
Media bottle,
with cap, round transparent borosilicate glass bottle, bottle capacity (500 mL), non-sterile
Expand
View Pricing
DWK14395-1000
KIMBLE
®
KIMAX
®
Media bottle,
with cap, round transparent borosilicate glass bottle, bottle capacity (1,000 mL), non-sterile
Expand
View Pricing
DWK14395-250
KIMBLE
®
KIMAX
®
Media bottle,
with cap, round transparent borosilicate glass bottle, bottle capacity (250 mL), non-sterile
Expand
View Pricing
DWK14395-100
KIMBLE
®
KIMAX
®
Media bottle,
with cap, round transparent borosilicate glass bottle, bottle capacity (100 mL), non-sterile
Expand
View Pricing
DWK14396-2000
KIMBLE
®
KIMAX
®
Media bottle,
without cap, round transparent borosilicate glass bottle, bottle capacity (2,000 mL), non-sterile
Expand
View Pricing
DWK14396-100
KIMBLE
®
KIMAX
®
Media bottle,
without cap, round transparent borosilicate glass bottle, bottle capacity (100 mL), non-sterile
Expand
View Pricing
DWK14396-1000
KIMBLE
®
KIMAX
®
Media bottle,
without cap, round transparent borosilicate glass bottle, bottle capacity (1,000 mL), non-sterile
Expand
View Pricing
DWK14396-10000
KIMBLE
®
KIMAX
®
Media bottle,
without cap, round transparent borosilicate glass bottle, bottle capacity (10,000 mL), non-sterile
Expand
View Pricing
DWK14396-5000
KIMBLE
®
KIMAX
®
Media bottle,
without cap, round transparent borosilicate glass bottle, bottle capacity (5,000 mL), non-sterile
Expand
View Pricing
DWK14396-500
KIMBLE
®
KIMAX
®
Media bottle,
without cap, round translucent borosilicate glass bottle, bottle capacity (500 mL), non-sterile
Expand
View Pricing
DWK14396-250
KIMBLE
®
KIMAX
®
Media bottle,
without cap, round transparent borosilicate glass, bottle capacity (250 mL), non-sterile
Expand
View Pricing