Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KIMBLE
®
disposable rimless culture tube
Products
Product Category
tubes (11)
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
DWK73500-1275
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (12 mm) (75 mm), non-sterile
Expand
View Pricing
DWK73500-650
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (6 mm) (50 mm), non-sterile
Expand
View Pricing
DWK73500-16100
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (16 mm) (100 mm), non-sterile
Expand
View Pricing
DWK73500-25150
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (25 mm) (150 mm), non-sterile
Expand
View Pricing
DWK73500-18150
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (18 mm) (150 mm), non-sterile
Expand
View Pricing
DWK60T1685B
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (16 mm) (85 mm), non-sterile
Expand
View Pricing
DWK73500-16150
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (16 mm) (150 mm), non-sterile
Expand
View Pricing
DWK73500-16125
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (16 mm) (125 mm), non-sterile
Expand
View Pricing
DWK73500-1075
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (10 mm) (75 mm), non-sterile
Expand
View Pricing
DWK73500-1585
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (15 mm) (85 mm), non-sterile
Expand
View Pricing
DWK73500-20150
KIMBLE
®
disposable rimless culture tube,
round bottom borosilicate glass, tube diam. (20 mm) (150 mm), non-sterile
Expand
View Pricing