Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kern pocket balances
Products
Product Category
balances (4)
Manufacturer
Height (mm)
Length (mm)
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z662682
Kern pocket balances,
weighing capacity 1,000 g, resolution: 1 g
Expand
View Pricing
Z662666
Kern pocket balances,
weighing capacity 150 g, resolution: 0.1 g
Expand
View Pricing
Z662674
Kern pocket balances,
weighing capacity 320 g, resolution: 0.1 g
Expand
View Pricing
Z662658
Kern pocket balances,
weighing capacity 60 g, resolution: 0.01 g
Expand
View Pricing