Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kern ABT analytical balances
Synonym(s):
Kern ABT analytical balances
Products
Product Category
balances (10)
Manufacturer
Diameter (mm)
Weighing Capacity (g)
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z757284
Kern ABT analytical balances,
model 220-4M, weighing capacity 220 g, AC/DC input 220 - 240 V AC, universal plug set
Expand
View Pricing
Z757292
Kern ABT analytical balances,
model 120-5DM, weighing capacity 42-120 g, AC/DC input 220 - 240 V AC, universal plug set
Expand
View Pricing
Z742901
Kern ABT analytical balances,
Kern ABT 100-5NM, weighing capacity 101 g, accuracy: 0.01 g, precision: ±0.15 g
Expand
View Pricing
Z757306
Kern ABT analytical balances,
model 220-5DM, weighing capacity 82-220 g, AC/DC input 220 - 240 V AC, universal plug set
Expand
View Pricing
Z757276
Kern ABT analytical balances,
model 120-4M, weighing capacity 120 g, AC/DC input 220 - 240 V AC, universal plug set
Expand
View Pricing
Z742903
Kern ABT analytical balances,
Kern ABT 320-4NM, weighing capacity 302 g, accuracy: 0.1 g, precision: ±0.3 g
Expand
View Pricing
Z674796
Kern ABT analytical balances,
weighing capacity 220 g, resolution: 0.1 mg, linearity: ±0.2 mg
Expand
View Pricing
Z674826
Kern ABT analytical balances,
weighing capacity 82 or 220 g, resolution: 0.01 or 0.1 mg, linearity: ±0.1 or 0.2 mg
Expand
View Pricing
Z674788
Kern ABT analytical balances,
weighing capacity 120 g, resolution: 0.1 mg, linearity: ±0.2 mg
Expand
View Pricing
Z674818
Kern ABT analytical balances,
weighing capacity 42 or 120 g, resolution: 0.01 or 0.1 mg, linearity: ±0.03 or 0.2 mg
Expand
View Pricing