Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank
Products
Product Category
baths (11)
Manufacturer
Capacity (L)
Color
Package Size
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z742956
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
BT9, transparent plastic bath, capacity 6-9 L
Expand
View Pricing
Z742964
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
B27, stainless steel bath (Insulated), capacity 17 - 27 L
Expand
View Pricing
Z742955
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
BT5, transparent plastic bath, capacity 3.5 - 5 L
Expand
View Pricing
Z742958
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
BT27, transparent plastic bath, capacity 20 - 27 L
Expand
View Pricing
Z742965
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
B33, stainless steel bath (Insulated), capacity 26 - 39 L
Expand
View Pricing
Z742957
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
BT19, transparent plastic bath, capacity 14 - 19 L
Expand
View Pricing
Z742966
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
B39, stainless steel bath (Insulated), capacity 35 - 41 L
Expand
View Pricing
Z742959
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
B5, stainless steel bath (Insulated), capacity 3.5 - 5 L
Expand
View Pricing
Z742963
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
B19, stainless steel bath (Insulated), capacity 14 - 19 L
Expand
View Pricing
Z742960
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
B13, stainless steel bath (Insulated), capacity 9 - 13 L
Expand
View Pricing
Z742962
Julabo
®
CORIO
™
C/CD heating immersion circulator bath tank,
B17, stainless steel bath (Insulated), capacity 13 - 17 L
Expand
View Pricing