Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron/nickel
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
GF49707669
Iron/nickel,
sheet, Fe 52%/Ni 48%, thickness 7.0 mm, size 108 × 108 mm, temper annealed
Expand
View Pricing
GF54256507
Iron/nickel,
powder, Fe 58%/Ni 42%, max. particle size 150 micron, weight 100 g, condition alloy pre-cursor
Expand
View Pricing
GF83913655
Iron/nickel,
powder, 45 max. part. size (micron), Fe58%/Ni42%, weight 50 g, condition atomized
Expand
View Pricing
GF58365041
Iron/nickel,
rod, Fe 55%/Ni 45%, 16 mm diameter, length 100 mm, temper unannealed
Expand
View Pricing
GF82803517
Iron/nickel,
powder, 45 max. part. size (micron), Fe58%/Ni42%, weight 100 g, condition atomized
Expand
View Pricing
GF47942466
Iron/nickel,
rod, Fe 54%/Ni 46%, 8.2 mm diameter, length 100 mm, temper as drawn
Expand
View Pricing
GF20924197
Iron/nickel,
rod, Fe 54%/Ni 46%, 8.2 mm diameter, length 200 mm, temper as drawn
Expand
View Pricing
GF80790240
Iron/nickel,
rod, Fe54%/Ni46%, 8.2 mm diameter, length 1000 mm, temper as drawn
Expand
View Pricing
GF51505264
Iron/nickel,
rod, Fe 55%/Ni 45%, 16 mm diameter, length 50 mm, temper unannealed
Expand
View Pricing
GF82063032
Iron/nickel,
powder, 150 max. part. size (micron), Fe58%/Ni42%, weight 200 g, condition alloy pre-cursor
Expand
View Pricing
GF93309161
Iron/nickel,
powder, Fe58%/Ni42%, 45 max. part. size (micron), weight 200 g, condition atomized
Expand
View Pricing
GF12107887
Iron/nickel,
rod, Fe 55%/Ni 45%, 16 mm diameter, length 200 mm, temper unannealed
Expand
View Pricing
GF45895326
Iron/nickel,
rod, Fe 54%/Ni 46%, 8.2 mm diameter, length 500 mm, temper as drawn
Expand
View Pricing
GF81234053
Iron/nickel,
powder, Fe58%/Ni42%, 150 max. part. size (micron), weight 50 g, condition alloy pre-cursor
Expand
View Pricing