Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron/boron/silicon
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-20 of 33
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF16506817
Iron/boron/silicon,
foil, Fe 78%/B 13%/Si 9%, thickness 0.025 mm, length 0.5 m, coil width 50 mm
Expand
View Pricing
GF17690733
Iron/boron/silicon,
wire, Fe 77.5%/B 15%/Si 7.5%, 0.05 mm diameter, length 2 m, condition metallic glass
Expand
View Pricing
GF18952959
Iron/boron/silicon,
foil, Fe 78%/B 13%/Si 9%, thickness 0.025 mm, length 2 m, coil width 50 mm
Expand
View Pricing
GF38499104
Iron/boron/silicon,
foil, Fe 79%/B 16%/Si 5%, thickness 0.025 mm, length 0.5 m, coil width 50 mm
Expand
View Pricing
GF89658837
Iron/boron/silicon,
foil, Fe81%/B 13.5%/Si 3.5%/C 2%, thickness 0.025 mm, length 2 m, coil width 50 mm
Expand
View Pricing
GF63351626
Iron/boron/silicon,
foil, Fe78%/B 13%/Si 9%, thickness 0.025 mm, length 1 m, coil width 50 mm
Expand
View Pricing
GF68946036
Iron/boron/silicon,
wire, Fe77.5%/B 15%/Si 7.5%, length 20 m, 0.02 mm diameter, condition metallic glass
Expand
View Pricing
GF74951567
Iron/boron/silicon,
wire, Fe77.5%/B 15%/Si 7.5%, length 10 m, 0.02 mm diameter, condition metallic glass
Expand
View Pricing
GF11342483
Iron/boron/silicon,
wire, Fe 77.5%/B 15%/Si 7.5%, 0.05 mm diameter, length 5 m, condition metallic glass
Expand
View Pricing
GF12387540
Iron/boron/silicon,
foil, Fe 79%/B 16%/Si 5%, thickness 0.025 mm, length 1 m, coil width 50 mm
Expand
View Pricing
GF39868060
Iron/boron/silicon,
foil, Fe 78%/B 13%/Si 9%, thickness 0.025 mm, length 10 m, coil width 50 mm
Expand
View Pricing
GF68182285
Iron/boron/silicon,
wire, Fe77.5%/B 15%/Si 7.5%, 0.125 mm diameter, condition metallic glass, length 50 m
Expand
View Pricing
GF85572366
Iron/boron/silicon,
wire, Fe77.5%/B 15%/Si 7.5%, 0.125 mm diameter, length 10 m, condition metallic glass
Expand
View Pricing
GF07354223
Iron/boron/silicon,
foil, Fe 81%/B 13.5%/Si 3.5%/C 2%, thickness 0.025 mm, length 5 m, coil width 50 mm
Expand
View Pricing
GF26380739
Iron/boron/silicon,
wire, Fe 77.5%/B 15%/Si 7.5%, 0.05 mm diameter, length 20 m, condition metallic glass
Expand
View Pricing
GF27007332
Iron/boron/silicon,
foil, Fe 81%/B 13.5%/Si 3.5%/C 2%, thickness 0.025 mm, length 0.5 m, coil width 50 mm
Expand
View Pricing
GF30469642
Iron/boron/silicon,
foil, Fe 79%/B 16%/Si 5%, thickness 0.025 mm, length 10 m, coil width 50 mm
Expand
View Pricing
GF48443553
Iron/boron/silicon,
foil, Fe 79%/B 16%/Si 5%, thickness 0.025 mm, length 2 m, coil width 50 mm
Expand
View Pricing
GF69118948
Iron/boron/silicon,
foil, thickness 0.025 mm, Fe78%/B 13%/Si 9%, length 5 m, coil width 50 mm
Expand
View Pricing
GF95262396
Iron/boron/silicon,
wire, Fe77.5%/B 15%/Si 7.5%, 0.02 mm diameter, length 2 m, condition metallic glass
Expand
View Pricing
Page 1 of 2
Page 1 of 2