Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Inter-Seal
™
bonded caps (inseparable caps and liner)
Products
Product Category
caps (2)
Brand
Supelco (2)
Color
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SU860006
Inter-Seal
™
bonded caps (inseparable caps and liner),
white polypropylene cap (solid), tan PTFE/silicone (EPA Quality), thickness 3.2 mm, thread for 24-400, for use with 20 or 40mL vial, pkg of 100 ea
Expand
View Pricing
SU860005
Inter-Seal
™
bonded caps (inseparable caps and liner),
white polypropylene cap (15mm center hole), tan PTFE/silicone, thickness 3.2 mm, thread for 24-400, for use with 20 or 40 mL vial, pkg of 1000 ea
Expand
View Pricing