Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Inositol
Synonym(s):
myo
-Inositol, 1,2,3,4,5,6-Hexahydroxycyclohexane,
i
-Inositol,
meso
-Inositol
Empirical Formula (Hill Notation):
C
6
H
12
O
6
CAS Number:
87-89-8
Molecular Weight:
180.16
EC Number:
201-781-2
Products
Brand
Supelco (1)
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1340960
Inositol,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR1351
Inositol,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing