Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Infrared PbS quantum dots
Linear Formula:
PbS
CAS Number:
1314-87-0
Products
Product Category
quantum dots (1)
Brand
Sigma-Aldrich (6)
Physical Form
Quality Segment
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
925535
Infrared PbS quantum dots,
λ
max
, 1550 nm, 100 mg/mL in toluene
Expand
View Pricing
925543
Infrared PbS quantum dots,
λ
max
, 1350 nm, 100 mg/mL in toluene
Expand
View Pricing
935093
Infrared PbS quantum dots,
λ
max
, 1250 nm, 100 mg/mL in toluene
Expand
View Pricing
935115
Infrared PbS quantum dots,
λ
max
, 2000 nm, 100 mg/mL (in toluene)
Expand
View Pricing
935085
Infrared PbS quantum dots,
λ
max
, 1650 nm, 100 mg/mL in toluene
Expand
View Pricing
935107
Infrared PbS quantum dots,
λ
max
, 1850 nm, 100 mg/mL in toluene
Expand
View Pricing